Kimtech™ Sterling™ 9.5 in. Nitrile Gloves
Actual product may vary
Kimtech™ Sterling™ 9.5 in. Nitrile Gloves
Sterling™ gloves combine the sensitivity of latex with the protection and strength of nitrile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC50706Small
KLC50705X-Small
FIS19-048-132Small
KLC50707Medium
KLC50708Large
Catalog number KLC50706
Price (SGD)
480.00
Special offer
Ends: 06-Sep-2026
578.00
Save 98.00 (17%)
Case
Size:
Small
Price (SGD)
480.00
Special offer
Ends: 06-Sep-2026
578.00
Save 98.00 (17%)
Case
Sterling™ gloves combine the sensitivity of latex with the protection and strength of nitrile.
  • Standard 9.5 in. length
  • Powder free
  • Non-sterile
  • Static dissipative
  • Textured fingertips
  • Beaded cuff
  • Ambidextrous
  • Typical uses include: Healthcare, Laboratory, Cleaning, Maintenance, Electronic Assembly and Inspection
Specifications
Certifications/ComplianceASTM D 6319, Meets USP <800> guidelines
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory
AQL1
ColorGray
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)3.5 mil
Finger Thickness (Metric)0.08 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.1 mil
Palm Thickness (Metric)0.07 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeSmall
SterilityNon-sterile
Thickness (English)3.5 mil
Thickness (Metric)0.08 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
Model50706
Quantity1700 Gloves/Cs.
SeriesKimtech™
Tensile Strength42 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizeCase