L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure
Thermo Scientific Chemicals
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure
CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSKJ16215.C425 kgPlastic Bottle
Catalog number FSKJ16215.C4
Price (SGD)
1,846.00
Each
Quantity:
25 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
1,846.00
Each
Specifications
Chemical Name or MaterialL-Glutamic acid
Melting Point189°C to 192°C (decomposition)
CAS56-86-0
Health Hazard 1H500