Solutions A+ Aloe Latex Gloves
Actual product may vary
Fisherbrand
Solutions A+ Aloe Latex Gloves
Protects hands from drying, chapping and cracking. Fisherbrand™ Solutions A+ Aloe Latex Gloves combine protection and comfort.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS15173269Medium
FKS15163269Large
FKS15153269Small
Catalog number FKS15173269
Price (SGD)
-
Size:
Medium
Latex gloves provide a protective barrier and contain soothing Aloe vera to help prevent skin from drying, cracking and chapping. Textured to provide excellent grip. Powder free. EN 374:2003 (AQL 1.5).
  • PPE Category III
  • Disposable
  • Aloe vera coating protects from drying, chapping and cracking
  • Ideal for prolonged use
  • Textured surface provides good grip
  • Latex
Specifications
Certifications/ComplianceISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved
ColorNatural
HandAmbidextrous
MaterialLatex
PowderedPowder Free
SizeMedium
SterilityNon-sterile
Thickness (English)5.5 mil
For Use With (Application)Ideal for workers who have frequent glove changes
Glove Cuff StyleBeaded/Rolled
Quantity100/Pk.
Tensile Strength18 MPa
Length (Metric)240 mm
Unit SizePack