MagLab™ MagRack™ Tube Rack
MagLab™ MagRack™ Tube Rack
Actual product may vary
Fisherbrand
MagLab™ MagRack™ Tube Rack
Store centrifuge tubes without crowding limited workspace. Fisherbrand™ MagLab™ MagRack™ Tube Rack uses magnetic mounting to store 15mL and 50mL tubes off the benchtop. Constructed of durable, teal-colored ABS plastic.
Have Questions?
Catalog NumberHeightMetric
FKS11982581
Catalog number FKS11982581
Price (SGD)
-
Create lab storage space off your bench top with the MagLab™ storage system
  • Easily holds four 50mL tubes and nine 15mL tubes off your benchtop
  • Robust ABS plastic is translucent for easy content identification and access
  • Features four powerful magnets for a secure hold onto most metal surfaces
  • Mount on non-metallic surfaces with a self-adhesive metal mounting plate, provided
Specifications
Product TypeTube Rack
ColorTranslucent Teal
HoldsTubes (4 x 50 mL and 9 x 15 mL)
MaterialABS Plastic
Positions13
For Use With (Equipment)15mL and 50mL Centrifuge Tubes
No. per Pack1 Each
Quantity1 Each
Dimensions (L x W x H)131 x 112 x 64mm
Length (Metric)127 mm
Width (Metric)131 mm
Height (Metric)64 mm
Unit SizeEA