Immuno Breakable Modules Clear
Actual product may vary
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Thermo Scientific
Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentWell ShapeNo. per PackProduct Type
FIS22-047027PolySorpRound, C Bottom5Strip Plate
446469MaxiSorpRound, C Bottom10Strip Plate
446639MaxiSorpRound10Microplate Strip
Catalog number FIS22-047027
Price (SGD)
2,100.00
Case
Surface Treatment:
PolySorp
Well Shape:
Round, C Bottom
No. per Pack:
5
Product Type:
Strip Plate
Price (SGD)
2,100.00
Case
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
  • Free-standing modules are easy to handle
Specifications
BreakapartBreakable Strips (LockWell)
Binding PropertyHydrophobic
CertifiedNo
FormatStrip Plate
LidWithout Lid
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
Array8 x 12
PackagingInner packs of 5
ColorClear
Product TypeStrip Plate
MaterialPolystyrene
Volume (Metric) Well350 μL
No. of Wells96
Volume (Metric) Working250 μL
SterilityNon-sterile
Surface TreatmentPolySorp
Well ShapeRound, C Bottom
For Use With (Application)For adsorption of hydrophobic molecules
No. per Case60
No. per Pack5
Plate BlockingNone
Quantity60/Cs.
Target MoleculeLipids
Well DesignC8
Unit SizeCase