MICROFLEX™ Ultraform™ UF-524 Nitrile Gloves
Actual product may vary
Microflex
MICROFLEX™ Ultraform™ UF-524 Nitrile Gloves
Powder free, thin nitrile glove designed with ERGOFORM™ technology to support musculoskeletal health. Soft nitrile forms to hands for comfort during extended wear.
Have Questions?
Catalog NumberSize
FIS19-167-301X-Small
Catalog number FIS19-167-301
Price (SGD)
87.00
Pack
Size:
X-Small
Price (SGD)
87.00
Pack
  • Thin design for tactile sensitivity
  • Soft nitrile formulation forms to hands for comfort during extended wear
  • Cobalt blue color
  • Non-sterile
  • Ambidextrous
  • Textured fingertips
  • Beaded cuff
  • Ideal for a variety of markets including healthcare, life science and food processing
Specifications
Certifications/ComplianceASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe)EN 420:2003 + A1:2009, EN ISO 13485:2012
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory, Industrial
AQL1.5
ColorBlue
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)3.1 mil
Finger Thickness (Metric)0.08 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)2.4 mil
Palm Thickness (Metric)0.06 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeX-Small
SterilityNon-sterile
Thickness (English)2.4 mil
Thickness (Metric)0.06 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
ModelUF-524-XS
Quantity3000/Cs.
SeriesMICROFLEX™ Ultraform™ UF-524
Tensile Strength23 MPa
Length (English)9.1 in.
Length (Metric)23.1 cm
Unit SizePack