Radioactive Bags
Actual product may vary
Associated Bag
Radioactive Bags
Have Questions?
Catalog NumberMaterial
FIS19-120-2037LDPE, LLDPE
Catalog number FIS19-120-2037
Price (SGD)
674.00
Pack
Material:
LDPE, LLDPE
Price (SGD)
674.00
Pack
Specifications
Certifications/ComplianceFederal regulation 10-20.203
ColorYellow
Length (Metric)91.4 cm
Weight (English)44 lb.
Weight (Metric)19.9 kg
Width (Metric)61 cm
For Use With (Application)For safe disposal of radioactive waste
Thickness3 mm
MaterialLDPE, LLDPE
Unit SizePack