Coalescing Carbon Filters
Actual product may vary
Justrite
Coalescing Carbon Filters
Use with Justrite Quick-Disconnect Safety Cans
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS19-052-373Justrite Quick Disconnect Cans with SS Fittings
Catalog number FIS19-052-373
Price (SGD)
886.00
Each
For Use With (Equipment):
Justrite Quick Disconnect Cans with SS Fittings
Price (SGD)
886.00
Each
  • Ideal for HPLC applications—minimize odor by capturing 99% of liquid particulates released in the form of gaseous vapor
  • Two-stage vent filter is easy to install, snapping conveniently into factory installed quick-disconnect fitting
  • Color-changing carbon cartridge (top portion) changes from purple to black as hydrocarbons are absorbed, indicating when replacement is necessary
  • Coalescing filter (bottom portion) can be reused with new carbon filters several times and replaced after 3-4 carbon filter cycles
Cannot be used with 1-gal. oval style cans. Do not transport safety can while carbon filter is attached.
Specifications
Certifications/ComplianceEPA 40 CFR 264.173
Product TypeCoalescing Carbon Filter
For Use With (Application)HPLC
For Use With (Equipment)Justrite Quick Disconnect Cans with SS Fittings
MaterialPlastic
Width (English)4 in.
Width (Metric)10.16 cm
Height (English)11 in.
Height (Metric)27.94 cm
Unit SizeEach