Durx™ 770 Wipers
Actual product may vary
Berkshire
Durx™ 770 Wipers
For ISO Class 5 (Class 100) applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)
FIS19-0372594 in.
FIS06-665-33G9 in.
FIS17-987-79B7 in.
FIS17-987-80112 in.
FIS17-987-82018 in.
Catalog number FIS19-037259
Price (SGD)
42.00
Pack
Length (English):
4 in.
Price (SGD)
42.00
Pack
  • Made from mechanically hydroentangled 55% cellulose/45% polyester with proprietary non-ionic binder
  • Work well with IPA and other solvents
  • Low fiber and particle generation
  • Soft, highly sorbent and durable
  • Critical combination of synthetic polyester strength with the absorbent characteristics of cellulose
Specifications
Product TypeCleanroom Wipe
Length (English)4 in.
Length (Metric)10 cm
Width (English)4 in.
Width (Metric)10 cm
MaterialPolyester Cellulose
AbsorbancyHighly absorbent
CompatibilityCommon cleaning and disinfecting solutions
AutoclavableAutoclavable
Basis Weight65.8
Certifications/ComplianceRoHS
ColorWhite
Dimensions (L x W)4 x 4 in. (10.16 x 10.16 cm)
For Use With (Application)Cleaning of lab equipment, integrated circuit, printed circuit board and electronic components
Unit SizePack