Durx 570 Wiper
Actual product may vary
Berkshire
Durx 570 Wiper
For use in ISO Class 5 to 8 controlled environments
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-035-26922.86 x 22.86 cm (9 x 9 in.)
Catalog number FIS19-035-269
Price (SGD)
1,245.00
Case
Dimensions (L x W):
22.86 x 22.86 cm (9 x 9 in.)
Price (SGD)
1,245.00
Case
  • Autoclavable
  • Free of chemical additives
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)23 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialNon-woven Polyester Cellulose
Dimensions (L x W)22.86 x 22.86 cm (9 x 9 in.)
For Use With (Application)Designed for use in ISO Class 5 and higher cleanroom environments and USP <797> applications