▸
Racks
▸
Special Purpose Racks
▸
Stomacher™ Bag Racks
Actual product may vary
Seward
Stomacher™ Bag Racks
Seward has developed a full range of Stomacher™ Accessories to help provide a safe and efficient working environment in your laboratory.
Have Questions?
Catalog Number
Holds
For Use With (Equipment)
Dimensions (L x W x H)
FIS14-285-1
Bags
Stomacher 400 lab blenders
8 cm x 10.5 cm x 19 cm
Catalog number
FIS14-285-1
Price (SGD)
245.00
Each
Add to cart
Holds:
Bags
For Use With (Equipment):
Stomacher 400 lab blenders
Dimensions (L x W x H):
8 cm x 10.5 cm x 19 cm
Price (SGD)
245.00
Each
Add to cart
Help to organize sample bags
High-grade plastic polymer construction
Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
Perfect for loading closure bags pre-enrichment into the incubator
Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Specifications
Autoclavable
Not Autoclavable
Color
Clear
Holds
Bags
Material
Polymer
Positions
1
Sterility
Non-sterile
For Use With (Application)
To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment)
Stomacher 400 lab blenders
No. per Pack
1
Quantity
1
Dimensions (L x W x H)
8 cm x 10.5 cm x 19 cm
Weight (Metric)
0.4kg
Product Type
Bag Rack