Storage Microplates
Actual product may vary
Axygen
Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Have Questions?
Catalog NumberColor
FIS14-222-354Clear
Catalog number FIS14-222-354
Price (SGD)
118.00
Pack
Color:
Clear
Price (SGD)
118.00
Pack
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
  • 2 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications
    Certifications/ComplianceANSI, SLAS 1-2004
    FormatStandard
    LidWithout Lid
    Array8 x 12
    AutoclavableAutoclavable
    ColorClear
    Product Type96 Round Deep-Well Storage Microplate
    MaterialPolypropylene
    Volume (Metric) Well1.8 mL
    No. of Wells96
    SterilitySterile
    Well ShapeRound
    For Use With (Application)Sample collection, in-vitro growth chambers and long term storage
    Max. RCF4000xG
    No. per Case50/Cs.
    No. per Pack5/Pk.
    Temperature Range (English)-112°F
    Temperature Range (Metric)-80°C
    Unit SizePack