Storage Microplates
Actual product may vary
Axygen
Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityNo. of WellsVolume (Metric) Well
FIS14-222-353ClearNon-sterile961.8 mL
FIS14-222-231ClearSterile485000 μL
FIS14-222-352ClearSterile961100 μL
FIS14-222-350ClearSterile2410,000 μL
Catalog number FIS14-222-353
Price (SGD)
96.00
Pack
Color:
Clear
Sterility:
Non-sterile
No. of Wells:
96
Volume (Metric) Well:
1.8 mL
Price (SGD)
96.00
Pack
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications
    Certifications/ComplianceANSI, SLAS 1-2004
    FormatStandard
    LidWithout Lid
    Array8 x 12
    AutoclavableAutoclavable
    ColorClear
    MaterialPolypropylene
    Volume (Metric) Well1.8 mL
    No. of Wells96
    SterilityNon-sterile
    Well ShapeRound
    For Use With (Application)Sample collection, in-vitro growth chambers and long term storage
    Max. RCF4000 x g
    No. per Case50/Cs.
    No. per Pack5/Pk.
    Temperature Range (English)-112°F
    Temperature Range (Metric)-80°C
    Unit SizePack