96-well PCR Microplates
Actual product may vary
96-well PCR Microplates
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSkirt StyleFormat
FIS14-222-326ClearFullStandard
FIS14-222-320ClearElevatedStandard
FIS14-222-340ClearHalfSingle Notch Corner
Catalog number FIS14-222-326
Price (SGD)
154.00
Pack
Color:
Clear
Skirt Style:
Full
Format:
Standard
Price (SGD)
154.00
Pack
Axygen™ 96 well PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • Well Volume: 200 μL
Specifications
BarcodeNo Barcode
Certifications/ComplianceRNase-, DNase-free
FormatStandard
LidWithout Lid
Array8 x 12
ColorClear
For Use With (Equipment)Standard 96 well blocks
Skirt StyleFull
MaterialPolypropylene
Volume (Metric) Well200 μL
No. of Wells96
SterilityNon-sterile
Well ShapeRound
For Use With (Application)PCR thermal transfer, RT-PCR applications
No. per Case50/Cs.
No. per Pack10/Pk.
Unit SizePack