MICROFLEX™ Diamond Grip™ MF-300 Latex Gloves
Actual product may vary
Microflex
MICROFLEX™ Diamond Grip™ MF-300 Latex Gloves
Powder free, latex gloves are extra thick at the fingers for added protection against rips, snags and tears. Textured fingers help wearers securely grip tools and materials in a variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS11-462-67DLarge
FIS11-462-67AX-Small
FIS11-462-67BSmall
FIS11-462-67CMedium
Catalog number FIS11-462-67D
Price (SGD)
49.00
Pack
Size:
Large
Price (SGD)
49.00
Pack
Perfect choice for procedures that require greater tactile sensitivity for manipulation.
  • Made of premium quality, low modulus latex
  • Offer high-tensile strength for excellent resiliency
  • Thicker than the typical latex exam glove
  • Textured surface ensures secure grip
  • Beaded cuff for easy donning
  • Ambidextrous, nonsterile, single-use
  • Length: 9.6 in. (24.4cm)
  • Thickness: 6.3mil
  • Natural color
Specifications
Certifications/ComplianceASTM D3578, FDA21 CFR 177-2600
Certifications/Compliance (Europe)EN 420:2003 + A1:2009
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory, Industrial
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)7.9 mil
Finger Thickness (Metric)0.20 mm
Laboratory/Cleanroom UseNo
Latex FreeNo
MaterialLatex
Palm Thickness (English)6.3 mil
Palm Thickness (Metric)0.16 mm
PowderedPowder Free
Product TypeLatex Examination Gloves
SizeLarge
SterilityNon-sterile
Thickness (English)7.9 mil
Thickness (Metric)0.20 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
ModelMF-300-L
SeriesDiamond Grip™ MF-300
Tensile Strength30 MPa
Length (English)9.6 in.
Length (Metric)24.4 cm
Product LineMICROFLEX Diamond Grip
Unit SizePack