Drying Chamber for FreeZone™ Freeze-Dry Systems
Actual product may vary
Labconco
Drying Chamber for FreeZone™ Freeze-Dry Systems
Drying Chamber for use on FreeZone™ Freeze-Dry Systems. 12-port for 1, 2.5, 6, 12, and 18 L models; 16-port for 6, 12, and 18 L models.
Have Questions?
Catalog NumberNo. of PortsDiameter (English)
FIS10-273-19128 in.
Catalog number FIS10-273-19
Price (SGD)
9,905.00
Each
No. of Ports:
12
Diameter (English):
8 in.
Price (SGD)
9,905.00
Each
For use on FreeZone™ Freeze-Dry Systems: 1, 2.5, 4.5, 6, 8, 12 or 18 Liter models
  • Place small sample inside freeze-dry flasks and attach to valves
  • Type 304 stainless-steel construction
  • Weld-free for dependable vacuum operation
  • Neoprene valves accommodate both 0.5 and 0.75 in. adapters, gaskets and clear acrylic top for complete visibility during bulk drying
  • Compatible with FreeZone 1, 2.5, 4.5, 6, 8, 12 or 18 Liter Freeze Dryers
Specifications
TypeDrying Chamber
No. of Shelves0
MaterialStainless Steel
No. of Ports12
For Use With (Application)To provide a means to attach flasks to freeze dry system or contain samples in bulk
For Use With (Equipment)FreeZone 1, 2.5, 4.5, 6, 8, 12 or 18 Liter Freeze Dryers.
Includes12 neoprene valves
Diameter (English)8 in.
Diameter (Metric)20 cm
Height (English)9.75 in.
Height (Metric)24.7 cm
Weight (English)11 lb.
Weight (Metric)5 kg
Unit SizeEach