Panasonic™ 1000 Watt Commercial Microwave Oven
Actual product may vary
CurranTaylor
Panasonic™ 1000 Watt Commercial Microwave Oven
Perfect for heating of samples, general lab use, breakrooms and food prep areas
Have Questions?
Catalog NumberAmperage
FIS09-540-12713.4 A
Catalog number FIS09-540-127
Price (SGD)
3,033.00
Each
Amperage:
13.4 A
Price (SGD)
3,033.00
Each
  • Stainless steel front, steel exterior and cavity with ceramic shelf
  • Bottom energy feed
  • 10 programmable memory pads
  • Braille keypad
  • 20-memory capability
  • Double quantity key
  • 6 power levels
  • 2- and 3-stage cooking
  • Programmable lock
  • Unique “quick pick” preset times
  • Self-diagnostics
  • Output power 1000 Watts† 
For product datasheet, request BN20201723 from your Fisher Scientific Sales Rep.
Specifications
Certifications/ComplianceUL, DHHS, FCC, NSF, ADA
TypeCommercial Microwave Oven
IncludesStainless steel front, steel exterior and cavity with ceramic shelf, 10 programmable memory pads, braille keypad, double quantity key
Plug TypeNEMA 5-15R or NEMA 5-20R
Timer99 min 99 sec
Width (English)20.12 in.
Width (English) Interior13 in.
Height (English)12 in.
Height (English) Interior8 in.
Height (Metric)30.48 cm
Height (Metric) Interior20.32 cm
Depth (English) Interior13 in.
Depth (Metric) Interior33.02 cm
Shipping Weight (English)39 lb.
Shipping Weight (Metric)17.6 kg
Electrical Requirements120 V 60 Hz
Voltage120 V
Wattage1000 w
Amperage13.4 A
Frequency60 Hz
Unit SizeEach