Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Sterile and nonpyrogenic microplates with low evaporation lids.
Have Questions?
Catalog NumberPackaging
FIS07-200-87Bulk, 10 Bags x 5 Plates
Catalog number FIS07-200-87
Price (SGD)
917.00
Case
Packaging:
Bulk, 10 Bags x 5 Plates
Price (SGD)
917.00
Case
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Specifications
FormatStandard
LidWith Lid
Culture Area0.32 cm2
Array8 x 12
PackagingBulk, 10 Bags x 5 Plates
Binding TypeTC-Treated
ColorClear
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working0.1 to 0.2 mL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well0.36 mL
Well ShapeFlat Bottom (F-well)
No. per Pack50
Quantity50/Cs.
Diameter (Metric) Well0.64 cm
Unit SizeCase