MASTERBLOCK™ 96 Deep Well Conical Bottom 2mL Storage Plate
Actual product may vary
Greiner Bio-One
MASTERBLOCK™ 96 Deep Well Conical Bottom 2mL Storage Plate
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 2mL Storage Plate is also suitable for storing sample material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityNo. per Pack
FIS07-000-873Sterile1
FIS07-000-122Non-sterile5
Catalog number FIS07-000-873
Price (SGD)
895.00
Case
Sterility:
Sterile
No. per Pack:
1
Price (SGD)
895.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
AutoclavableNot Autoclavable
Binding TypeNon Binding
ColorClear
MaterialPolypropylene
No. of Wells96
Volume (Metric) Working0.1 to 2.0 mL
SterilitySterile
Total Volume (Metric) Well2 mL
Well ShapeSquare, Conical Bottom (V-well)
Dnase FreeYes
Max. RCF4800 x g (swinging bucket rotor)
No. per Case50
No. per Pack1
RNase FreeYes
Shelf Life5 Years after month of production
Temperature Range (Metric)-196°C to +121°C
Well ColorClear
Unit SizeCase