UV-Star™ 96-well Microplates
Actual product may vary
Greiner Bio-One
UV-Star™ 96-well Microplates
Ideal for measurements of DNA / RNA and protein concentrations at 260 and 280nm, respectively. Greiner Bio-One UV-Star™ 96-Well Microplates are eliminate the use of expensive and fragile quartz glass for routine laboratory work.
Have Questions?
Catalog NumberColor
FIS07-000-629Black
Catalog number FIS07-000-629
Price (SGD)
1,931.00
Case
Color:
Black
Price (SGD)
1,931.00
Case
  • Coated with μClear film well bottom to ensure low background for more accurate measurements
  • Innovative molding technique eliminates possibility of film detachment
  • Supplied without lid
  • Chimney-well design helps reduce cross-contamination
  • DNA measurement at 260/280nm
  • Protein measurement at 280nm
Specifications
BreakapartNo
FormatStandard
LidWithout Lid
Array8 x 12
PackagingInner bags of 10
Binding TypeNon Binding
ColorBlack
MaterialCycloolefin Copolymer
Volume (Metric) Well25 to 340 μL
No. of Wells96
SterilityNon-sterile
Surface TreatmentUV-Star
Total Volume (Metric) Well392 μL
Well ShapeRound, Flat Bottom (F-well)
No. per Case40
No. per Pack10
Quantity40/Cs.
Well ColorBlack
Dimensions (L x W x H)127.8 x 85.5 x 14.4 mm
Unit SizeCase