96-well No Bottom Microplates
Actual product may vary
Greiner Bio-One
96-well No Bottom Microplates
Made of polystyrene. Greiner Bio-One 96-Well No Bottom Microplates are designed without bottoms.
Have Questions?
Catalog NumberNo. of Wells
FIS07-000-62696
Catalog number FIS07-000-626
Price (SGD)
411.00
Case
No. of Wells:
96
Price (SGD)
411.00
Case
  • Virgin polystyrene construction
  • Chimney-well design to help prevent cross-contamination
Specifications
FormatStandard
Array8 x 12
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Well ShapeNo Bottom
Quantity40/Cs.
Well DesignChimney
Unit SizeCase