UV-Star™ 384-well Microplate Flat Bottom Microplate
Actual product may vary
Greiner Bio-One
UV-Star™ 384-well Microplate Flat Bottom Microplate
Used for measuring protein concentration at 280nm.
Have Questions?
Catalog NumberColor
FIS07-000-406Clear
Catalog number FIS07-000-406
Price (SGD)
2,874.00
Each
Color:
Clear
Price (SGD)
2,874.00
Each
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Used for nucleic acid determinations at 260nm/280nm
  • DMSO-resistant
Specifications
LidWithout Lid
Array16 x 24
AutoclavableNot Autoclavable
ColorClear
MaterialCycloolefin Copolymer
No. of Wells384
Volume (Metric) Working15 to 110 μL
SterilityNon-sterile
Total Volume (Metric) Well131 μL
Well ShapeRounded Square
Dnase FreeYes
Max. RCF4800 x g (swinging bucket rotor)
No. per Case40
No. per Pack10
Quantity40/Cs.
RNase FreeYes
Temperature Range (Metric)-80° to +40°C
Unit SizeEach