Low Profile Polystyrene Microplate Lids
Actual product may vary
Fisherbrand
Low Profile Polystyrene Microplate Lids
For use with 384 well standard microplates. Greiner Bio-one Low Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
Have Questions?
Catalog NumberSterility
FIS07-000-291Sterile
Catalog number FIS07-000-291
Price (SGD)
538.00
Case
Sterility:
Sterile
Price (SGD)
538.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Specifications
ColorClear
MaterialPolystyrene
SterilitySterile
No. of Wells384
Height (Metric)6 mm
Condensation RingsNo
No. per Case200
Plate Compatibility384-well Standard Microplate
Unit SizeCase