CELLSTAR μClear™ 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Greiner Bio-One
CELLSTAR μClear™ 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FIS07-000-166Black, Clear Bottom
Catalog number FIS07-000-166
Price (SGD)
811.00
Case
Color:
Black, Clear Bottom
Price (SGD)
811.00
Case
  • 96 Well Polystyrene Cell Culture Microplates, black
  • Lid with condensation rings; lid that enables gas exchange with minimal evaporation
  • Improved cell adhesion through physical surface treatment
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic
  • Non-Cytotoxic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of the microplates for optimal microscopic examinations
  • Stackable
  • Consecutive lot numbering
  • Have a uniform footprint, which is conform to the recommendation of the American National Standards Institute (ANSI) (ANSI/SBS 1-2004)
  • The risk of contamination from sample material being carried over is minimized
  • µClear bottom
  • Well profile: F-bottom/chimney well (flat bottom) 
  • Growth area per well: 34 mm²
  • Working volume per well: 25µl - 340µl  
  • Sterile 
Specifications
CertifiedFree of human DNA and pyrogens. Contents non-cytotoxic
LidLid with Condensation Rings
Culture Area0.34 cm2
Array8 x 12
AutoclavableNot Autoclavable
Packaging8/Pk., 32/Cs.
ColorBlack, Clear Bottom
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working0.025 to 0.340 mL
SterilitySterile
Surface TreatmentCell Culture
Well ShapeFlat Bottom (F-well)
Dnase FreeYes
Max. RCF4800 x g (swinging bucket rotor)
No. per Case32
No. per Pack8
Quantity32/Cs.
RNase FreeYes
Shelf Life4 Years
Temperature Range (Metric)-20°C to +60°C
Dimensions (L x W x H)128 x 85 x 14 mm
Unit SizeCase