Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Microplates
▸
Storage Microplates
▸
96-well Non-treated Polypropylene Microplates
Actual product may vary
Greiner
96-well Non-treated Polypropylene Microplates
Features high chemical resistance. Greiner Bio-One 96-Well Non-treated Polypropylene Microplates is free of detectable DNase, Rnase and human DNA.
Have Questions?
Change view
Catalog Number
Color
Sterility
Well Shape
FIS07-000-153
Clear
Non-sterile
Conical Bottom (V-well)
FIS07-000-110
Black
Non-sterile
Flat Bottom (F-well)
2 Options
Catalog number
FIS07-000-153
Price (SGD)
644.00
Case
Add to cart
Color:
Clear
Sterility:
Non-sterile
Well Shape:
Conical Bottom (V-well)
Price (SGD)
644.00
Case
Add to cart
Non-Pyrogenic
Free of detectable DNase, RNase, human DNA
Material: Polypropylene
High chemical resistance
Sealable with adhesive films and heat sealer
Can be traced back to production through defined LOT nu
Specifications
Format
Standard
Lid
Without Lid
Array
8 x 12
Color
Clear
Material
Polypropylene
No. of Wells
96
Sterility
Non-sterile
Surface Treatment
Non-treated
Well Shape
Conical Bottom (V-well)
No. per Case
100
Quantity
100/Cs.
Well Design
Chimney