Gamma Wipe™ 67 Wipers
Actual product may vary
Berkshire
Gamma Wipe™ 67 Wipers
For use in ISO Class 5 (Class 100) applications. Smooth, highly absorbent and durable with low extractable levels.
Have Questions?
Catalog NumberDimensions (L x W)
FIS06-665-38B12 x 12 in.
Catalog number FIS06-665-38B
Price (SGD)
954.00
Case
Dimensions (L x W):
12 x 12 in.
Price (SGD)
954.00
Case
  • Made from hydroentangled 55% cellulose/45% polyester
  • Ideal in sterile core areas where microbial exclusion is demanded
  • High level of absorbency and low level of particle generation
  • Sterilized with gamma radiation
    • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
    • Chemically compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches and quarternary ammonium compounds
Specifications
Product TypeCleanroom Wipe
Length (English)12 in.
Length (Metric)30.48 cm
Width (English)12 in.
Width (Metric)30.48 cm
MaterialPolyester Cellulose
CompatibilityCommon cleaning and disinfecting solutions
Basis Weight72.1
Certifications/ComplianceRoHS 2002/05, REACH
Dimensions (L x W)12 x 12 in.
For Use With (Application)For use in ISO Class 4 and higher sterile environments where microbial exclusion is demanded, excellent for cleaning medical devices
For Use With (Equipment)For cleaning sterile laminar air flow units, barrier isolators and bio-containment units