Talboys™ Labjaws™ Slide Staining Rack
Actual product may vary
Talboys™ Labjaws™ Slide Staining Rack
Resists corrosion under normal use
Have Questions?
Catalog number FIS02-216-335
Price (SGD)
326.00
Each
Price (SGD)
326.00
Each
Resists corrosion under normal use
  • Stainless steel construction
  • Adjustable to fit trays or sinks up to 21 in. (53.3cm) inside
  • L x W: 23.74 x 3.5 in. (60.3 x 8.9cm)
Specifications
MaterialStainless Steel
ShapeRectangular
Dimensions (L x W)23.74 x 3.5 in. (60.3 x 8.9cm)
Length (Metric)60.3 cm
Length (English)23.74 in.
Width (English)3.5 in.
Product TypeSlide Staining Rack
Width (Metric)8.9 cm
Unit SizeEach