Carrying Racks Compartment
Actual product may vary
Nasco
Carrying Racks Compartment
For transporting and using sample-filled bags in laboratories.
Have Questions?
Catalog NumberArrayDimensions (L x W x H)
FIS01-812-5E12 Compartments (3 x 4)25.4 x 25.4 x 9 cm
Catalog number FIS01-812-5E
Price (SGD)
139.00
Each
Array:
12 Compartments (3 x 4)
Dimensions (L x W x H):
25.4 x 25.4 x 9 cm
Price (SGD)
139.00
Each
For transporting and using sample-filled bags in laboratories.
  • Autoclavable
  • 10 and 12 gauge steel
  • Electrostatically applied powder coat finish
  • Max. dry temperature 300°F and wet 500°F
  • Easy sample transport and storage
Specifications
AutoclavableAutoclavable
ColorBlue
HoldsBags
MaterialCoated Wire
Positions12
ShapeRectangular
StackableNo
SterilityNon-sterile
ModelB00751
Quantity1 Ea.
Dimensions (L x W x H)25.4 x 25.4 x 9 cm
Length (Metric)25.4 cm
Length (English)10 in.
Width (English)10 in.
Height (English)3.5 in.
Product TypeSample Bag Carrying Rack
ShelvesNone
Height (Metric)9 cm
Array12 Compartments (3 x 4)
Width (Metric)25.4 cm
Unit SizeEach