Search Fisher Scientific
| Catalog Number | Sterility | Volume (Metric) Well | Well Shape | No. of Wells |
|---|---|---|---|---|
| CRG3959 | Non-sterile | 1000 μL | Round | 96 |
| FIS07-200-700 | Sterile | 2000 μL | Conical Bottom (V-well) | 96 |
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.