Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Actual product may vary
Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
CRG3799Individually wrapped, 50/Cs.
FIS07-200-95Individually wrapped, 50/Cs.
FIS07-200-720ABag of 20; Cs. of 100
Catalog number CRG3799
Price (SGD)
122.00
Case
Packaging:
Individually wrapped, 50/Cs.
Price (SGD)
122.00
Case
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile
Specifications
BarcodeNo Barcode
FormatStandard
LidWith Lid
Culture Area0.32 cm2
Array8 x 12
PackagingIndividually wrapped, 50/Cs.
Binding TypeTC-Treated
ColorClear
Product TypeCell Culture Microplate
MaterialPolystyrene
Volume (Metric) Well330 μL
No. of Wells96
Volume (Metric) Working75 to 200 μL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well0.33 mL
Well ShapeRound Bottom (U-well)
For Use With (Application)Attachment and growth of anchorage-dependent cells
No. per Case50/Cs.
No. per Pack1/Pk.
Quantity50/Cs.
Well ColorClear
Diameter (Metric) Well0.64 cm
Unit SizeCase