Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. per PackNo. per Case
CRG3596Clear1/Pk.50/Cs.
FKS10792552Clear1/Pk.100/Cs.
FIS07-200-91Clear5/Pk.100/Cs.
FIS07-200-706Clear20/Pk.100/Cs.
Catalog number CRG3596
Price (SGD)
107.00
Case
Color:
Clear
No. per Pack:
1/Pk.
No. per Case:
50/Cs.
Price (SGD)
107.00
Case
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
BarcodeNo Barcode
FormatStandard
LidWith Lid
Culture Area0.32 cm2
Array8 x 12
Binding TypeTC-Treated
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working0.1 to 0.2 mL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well0.36 mL
Well ShapeFlat Bottom (F-well)
No. per Case50/Cs.
No. per Pack1/Pk.
Quantity50/Cs.
Well ColorClear
Diameter (Metric) Well0.64 cm
Unit SizeCase