▸
Microplates
▸
Cell Culture Microplates
▸
Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Sterile and nonpyrogenic microplates with low evaporation lids.
Have Questions?
Change view
Catalog Number
Packaging
CRG3595
Individually Wrapped
FIS07-200-89
Individually Wrapped
FKS10687551
Individually Wrapped
FIS07-200-87
Bulk, 10 Bags x 5 Plates
4 Options
Catalog number
CRG3595
Price (SGD)
154.00
Special offer
Ends:
06-Sep-2026
253.00
Save 99.00 (39%)
Case
Add to cart
Packaging:
Individually Wrapped
Price (SGD)
154.00
Special offer
Ends:
06-Sep-2026
253.00
Save 99.00 (39%)
Case
Add to cart
Cell culture microplates are sterile and nonpyrogenic.
Lids available where indicated
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Specifications
Format
Standard
Lid
With Lid
Culture Area
0.32 cm
2
Array
8 x 12
Packaging
Individually Wrapped
Binding Type
TC-Treated
Color
Clear
Material
Polystyrene
No. of Wells
96
Volume (Metric) Working
0.1 to 0.2 mL
Sterility
Sterile
Surface Treatment
Cell Culture
Total Volume (Metric) Well
0.36 mL
Well Shape
Flat Bottom (F-well)
No. per Pack
50
Quantity
50/Cs.
Diameter (Metric) Well
0.64 cm