96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Catalog NumberProduct TypeVolume (Metric) Working
CRG353072Microplate1.5 to 2.2 mL
Catalog number CRG353072
Price (SGD)
135.00
Special offer
Ends: 06-Sep-2026
222.00
Save 87.00 (39%)
Case
Product Type:
Microplate
Volume (Metric) Working:
1.5 to 2.2 mL
Price (SGD)
135.00
Special offer
Ends: 06-Sep-2026
222.00
Save 87.00 (39%)
Case
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
Specifications
FormatStandard
LidWith Lid
Culture Area0.32 cm2
Array8 x 12
Packaging1 Pk., 50 Cases
Binding TypeTC-Treated
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working1.5 to 2.2 mL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well0.37 mL
Well ShapeFlat Bottom (F-well)
No. per Case50/Cs.
No. per Pack1/Pk.
Quantity50/Cs.
Unit SizeCase