PROSAT™ Sterile™ Meltblown Polypropylene Wipes
Actual product may vary
PROSAT™ Sterile™ Meltblown Polypropylene Wipes
PROSAT™ Sterile™ Meltblown Polypropylene Wipes
Contec
PROSAT™ Sterile™ Meltblown Polypropylene Wipes
These sterile, polypropylene wipes are presaturated with 70% USP grade isopropyl alcohol and 30% deionized water, and provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Quantity
CNTPS-911EB30 Wipes/Pk., 48 Pk./Cs.
FIS18-999-47430 Wipes/Pk., 48 Pk./Cs.
Catalog number CNTPS-911EB
Price (SGD)
-
Packaging Quantity:
30 Wipes/Pk., 48 Pk./Cs.
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for steril
These PROSAT™ presaturated wipes are exceptionally clean and also free from additives of any kind. This particular meltblown polypropylene contains very low levels of sodium and other ions. PROSAT™ meltblown polypropylene wipes are ideal for wiping articles prior to pass through, cleaning process tools and other equipment, and general surface cleaning.
  • Low in particles and fibers, making it ideal for product contact areas
  • Presaturated wipes reduce solvent usage and VOC emissions and ensure consistent saturation of each wipe independent of the operator
  • Validated sterile via gamma irradiation to a 10-6 Sterility Assurance Level
  • Compatible with Grade A/B or ISO Class 5 through 8 environments
  • Wipes are provided in a resealable pouch for ease of use and convenience
Specifications
Product TypePre-saturated Wipe
Length (English)11 in.
Length (Metric)28 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialPolypropylene
Certifications/ComplianceISO Class 5 to 8
Dimensions (L x W)28 x 23 cm (11 x 9 in.)
For Use With (Application)Routine Equipment Cleaning in Sterile Environments
Packaging Quantity30 Wipes/Pk., 48 Pk./Cs.
Saturation Solution70% IPA, 30% DI Water