96-well Sapphire Microplates
Actual product may vary
96-well Sapphire Microplates
Features optimal heat transfer from thermoblock to reaction solution.
Have Questions?
Catalog NumberSkirt StyleNo. per Case
ASO6-522-01Non-skirt40
Catalog number ASO6-522-01
Price (SGD)
-
Skirt Style:
Non-skirt
No. per Case:
40
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Ultra thin polypropylene for optimal heat transfer
  • Are particularly suited for automation
  • Robot-friendly- for grippers, sufficient space for the application
Specifications
BreakapartNo
FormatStandard
LidWithout Lid
Array8 x 12
AutoclavableNot Autoclavable
PackagingInner bags of 10
Binding TypeNon Binding
ColorClear
Skirt StyleNon-skirt
MaterialPolypropylene
No. of Wells96
SterilityNon-sterile
Total Volume (Metric) Well200 μL
No. per Case40
No. per Pack10
Quantity100/Cs.
Well ColorClear
Unit SizeEach