Anthranilic acid, 98+%
Anthranilic acid, 98+%
Acros Organics
Anthranilic acid, 98+%
CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol
Have Questions?
Catalog NumberQuantity
104911000100 g
Catalog number 104911000
HSN 29224300
Price (SGD)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAnthranilic acid
Melting Point144.0°C to 148.0°C
CAS118-92-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Causes serious eye damage.