X4 and S4 Series Centrifuges
Performance, Power and Versatility
General Purpose Centrifuges with GreenCool™ Technology
Thermo Fisher Scientific is proud to introduce the new Thermo Scientific™ X and S Series 4L General Purpose Centrifuges, combining over 75 years of engineering excellence with GreenCool™ Technology and a natural, non-fluorinated refrigerant system designed to meet upcoming global F- gas regulations, including EU and US EPA requirements.
Powerful, Versatile Centrifugation
The X4 and S4 General Purpose Centrifuges deliver the performance and flexibility needed to support a wide range of laboratory applications, while embracing a more sustainable future. .
Performance and versatility
Maximize performance and flexibility with up to 18 rotor options, tool-free Auto-Lock™ rotor exchange in seconds, fixed-angle rotors exceeding 25,000 x  g, swinging bucket rotors over 7,000 x g, and –20°C cooling in refrigerated models.
Greener by design™
Greener by design™, the refrigerated Thermo Scientific™ X4 and S4 Series Centrifuges use GreenCool™ Technology with R290, a natural refrigerant with a GWP of just 3—up to 475× lower environmental impact than conventional refrigerants—and include an end-of-life recycling program in the US.
Enhanced Sustainability Across the Lifecycle
Manufactured in Thermo Fisher Scientific’s zero- waste Center of Excellence in Osterode am Harz, Germany, the X and S Series centrifuges are powered by 100% renewable electricity and fully compliant with RoHS and WEEE directives for environmentally responsible manufacturing. 
Fit-for-Purpose Solutions for Every
Customize your centrifuge to fit your lab with benchtop or floor-standing models, a choice of high-performance X Series with touchscreen interface or streamlined S Series for routine applications, available in both ventilated and refrigerated formats.
Enhanced Sustainability
GreenCool Technology and more
General Purpose X4 and S4 Centrifuges are powerful and versatile solutions designed to meet most application needs. They offer enhanced performance features unleashed by the power of green technology.
Browse our product list—Shop now!
Catalog # Name Size Price Qty
75007343 Each
10,306.00
Special offer
Ends 30-Jun-2026
14,722.00 Save 4,416.00 (30%)
75007366 Each
19,656.00
75007333 Each
7,997.00
Special offer
Ends 30-Jun-2026
11,424.00 Save 3,427.00 (30%)
75007358 Each
16,136.00
Related Videos
Have a product question? Please complete and submit the form below.
Confirmed Opt-In (Optional)
Your confidentiality is important to us and all personal information that you provide will be used by us in accordance with our Privacy Policy and you will have the ability to opt out of receiving future marketing information from us at any time.