Check Order Status
Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
For rotary vane pump DN 16 KF
Have Questions?
Catalog number
WEL330044
Price (SGD)
392.00
EA
Add to cart
Price (SGD)
392.00
EA
Add to cart
Vacuum concentrator kit includes:
KF16 flange adapter
1.5m PVC vacuum hose
Clamps, centring rings and adapters
Specifications
Description
DN 16 KF
For Use With (Equipment)
Rotary vane pump DN 16 KF