1000FR Flame Retardant Protective Coverall
Actual product may vary
1000FR Flame Retardant Protective Coverall
1000FR Flame Retardant Protective Coverall
1000FR Flame Retardant Protective Coverall
1000FR Flame Retardant Protective Coverall
1000FR Flame Retardant Protective Coverall
1000FR Flame Retardant Protective Coverall
Flame Retardant Protective Coverall with Attached Hood, Elastic Wrist, and Elastic Back.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
UTCDD410-XLX-Large
UTCDD410-2XL2X-Large
Catalog number UTCDD410-XL
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
393.00
Save 73.00 (19%)
Case
Size:
X-Large
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
393.00
Save 73.00 (19%)
Case
  • High Moisture Vapor Transmission rate keeps your people cool and comfortable.
  • Well-designed garment provides worker comfort and a lower cost-in-use.
  • Certified for protection against harmful particulates, light liquid spray and it offers limit flame spread protection which meets EN ISO 14116 Index 1.
Specifications
Product TypeFlame-resistant Coveralls
SizeX-Large
GenderUnisex
MaterialFR Anti-static SMS Fabric
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeAttached, Open Face
Seam TypeSealed
SterilityNon-sterile
ResistanceFlame, Dust, Liquid
Unit SizeCase