▸
Solvents
▸
Toluene
▸
Toluene (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Toluene (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFT3241
1 L
Glass Bottle
Catalog number
USFT3241
Price (SGD)
78.00
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
78.00
Each
Add to cart
Specifications
Chemical Name or Material
Toluene
Cleanup Symbol
Solvent
Melting Point
-95°C
CAS
108-88-3
ChemAlert Storage Symbol
Red
View more