▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Sodium Thiosulfate Pentahydrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Thiosulfate Pentahydrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Standardized at 25°C against NIST™ potassium dichromate
Have Questions?
Catalog Number
Quantity
Packaging
USFSS3681
1 L
Poly Bottle
Catalog number
USFSS3681
Price (SGD)
41.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
41.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Thiosulfate Pentahydrate Solution
Color
Colorless
Name Note
0.1N (N/10)
CAS
10102-17-7
ChemAlert Storage Symbol
Gray
View more