Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Sodium Hypochlorite Solution (4-6%/Laboratory), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hypochlorite Solution (4-6%/Laboratory), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFSS2901
1 L
Poly Bottle
Catalog number
USFSS2901
Price (SGD)
51.00
Bottle
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
51.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Sodium Hypochlorite Solution
Color
Yellow
Name Note
4-6%
CAS
7681-52-9
ChemAlert Storage Symbol