Check Order Status
Sodium Hydroxide Solution (5N/Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide Solution (5N/Certified), Fisher Chemical
A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
USFSS256500
500 mL
Poly Bottle
Catalog number
USFSS256500
Price (SGD)
54.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
54.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
12.0
Cleanup Symbol
Caustic
Name Note
5N
Boiling Point
100°C
View more