Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Bases
▸
Potassium Hydroxide
▸
Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFSP236500
500 mL
Poly Bottle
Catalog number
USFSP236500
Price (SGD)
39.00
Bottle
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
39.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Potassium Hydroxide Solution
pH
12.0
Cleanup Symbol
Caustic
Color
Colorless
Name Note
45% w/w
View more