▸
Bases
▸
Potassium Hydroxide
▸
Potassium Hydroxide Solution, 0.1N (N/10) (In Methanol/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Potassium Hydroxide Solution, 0.1N (N/10) (In Methanol/Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFSP2241
1 L
Glass Bottle
Catalog number
USFSP2241
Price (SGD)
67.00
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
67.00
Each
Add to cart
Specifications
Chemical Name or Material
Potassium Hydroxide Solution
pH
13
Cleanup Symbol
Caustic
Color
Colorless
Name Note
0.1N (N/10), in methanol
View more