▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
USFSB141500
500 mL
Catalog number
USFSB141500
Price (SGD)
45.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
45.00
Each
Add to cart
Chemical Identifiers
CAS
7447-40-7, 1310-73-2, 139-33-3, 7732-18-5, 10043-35-3
Specifications
Chemical Name or Material
Buffer Concentrate, pH 10.01
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
Color
Undesignated
CAS
1310-73-2
View more