▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Buffer Solution, pH 10.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Buffer Solution, pH 10.00 (Certified), Fisher Chemical™
Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate
Have Questions?
Change view
Quantity:
500 mL
1 L
10 L
Packaging:
View all
Poly Bottle
Catalog number
USFSB116500
Price (SGD)
44.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
44.00
Each
Add to cart
Chemical Identifiers
CAS
6381-92-6, 584-08-7, 1310-58-3, 7732-18-5, 12228-88-5
Specifications
Chemical Name or Material
Buffer Solution, pH 10.00
CAS
6381-92-6
ChemAlert Storage Symbol
Gray
Color
Colorless
Health Hazard 2
CAUTION!
View more