▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Buffer Concentrate, pH 7.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Buffer Concentrate, pH 7.00 (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
USFSB1091
1 L
Catalog number
USFSB1091
Price (SGD)
63.00
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
63.00
Each
Add to cart
Chemical Identifiers
CAS
64-02-8, 7732-18-5, 1310-58-3, 1310-73-2, 7664-38-2
Specifications
Chemical Name or Material
Buffer Concentrate, pH 7.00
CAS
64-02-8
ChemAlert Storage Symbol
Gray
CAS
7732-18-5
CAS
1310-58-3
View more