▸
Acids
▸
Inorganic Acids
▸
Hydrochloric Acid
▸
Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
USFSA56500
500 mL
Glass Bottle
Catalog number
USFSA56500
Price (SGD)
75.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
75.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid Solution, 6N
pH
1
Cleanup Symbol
Acid
CAS
7647-01-0
ChemAlert Storage Symbol