Check Order Status
Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFS472500
500 g
Poly Bottle
Catalog number
USFS472500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Phosphate Dibasic Dihydrate
CAS
10028-24-7
ChemAlert Storage Symbol
Gray
Color
White
Health Hazard 2
CAUTION!
View more